Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NanoXact Gold Nanospheres 1 mg/ml (Au) Polystyrene Surface, nanoComposix

Supplier: nanoComposix

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AUYH10-5M AUYH10-1M AUYH50-5M AUYH100-10M AUYH10-10M AUYH100-1M AUYH50-1M AUYH50-10M AUYH100-5M
103820-080EA 463.08 USD InStock
103820-080 103820-070 103820-082 103820-072 103820-074 103820-076 103820-066 103820-078 103820-068
NanoXact Gold Nanospheres 1 mg/ml (Au) Polystyrene Surface, nanoComposix
Beads Microcarriers
High quality gold nanoparticles. Displaceable citrate surface. Provided with full COA; great value.

  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Standard colloidal concentration (0.05 mg/ml, 50 ppm, 0.254 mm, near 1 OD [0.8 to 1.6 depending on size])
  • Citrate surface is stable and displaceable

Spherical gold nanoparticles, 1 mg/ml, Polystyrene surface. NanoXact materials are precisely engineered, unagglomerated nanospheres with a narrow size distributions (CVs <12% for 7 to 15 nm sizes, <15% for 20 to 100 nm sizes, and <18% for 5 nm size).

Each batch is extensively characterized to ensure it meets the highest quality standards so you can achieve reliable & reproducible results. Products are supplied with COA's that include batch-specific TEM, DLS, Zeta Potential, UV-Vis, & pH measurements.

Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.

Custom formulations and ISO13485/cGMP compliant materials can be provided on request.

Delivery information: Ships at ambient temperatures

Caution: DO NOT FREEZE. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty