Would you like to log in again?
NanoXact Gold Nanoshells 0.05 mg/ml Carboxyl sufrace, nanoComposix
Supplier: nanoComposix
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GSLN660-500M GSLN660-1L GSLN660-100M
103820-300EA 387.69 USD InStock
103820-300 103820-302 103820-306
NanoXact Gold Nanoshells 0.05 mg/ml Carboxyl sufrace, nanoComposix
Beads Microcarriers
Nanoparticle with a silica core and gold shell; high concentration with near-infrared absorbance
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration gold nanoshells (1 mg/ml gold basis concentration, ~25 OD)
  • Used for optical filters, sensors, and photothermal therapies
Spherical gold nanoshell nanoparticles have a 120 nm silica core with a ~20 nm outer gold shell. Peak absorbance at 660 nm. Provided at 0.05 mg/ml gold concentration. Used in optical filters, sensing, and photothermal therapies. The carboxyl (lipoic acid) surface provide useful functionalization for conjugation to antibodies.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty