Would you like to log in again?
NanoXact Magnetite Nanoparticles 20 mg/ml PVP (polyvinylpyrrolidone), nanoComposix
Supplier: nanoComposix
MGPN20-10M MGPN20-5M MGPN20-25M
103820-392EA 398.46 USD InStock
103820-392 103820-396 103820-398
NanoXact Magnetite Nanoparticles 20 mg/ml PVP (polyvinylpyrrolidone), nanoComposix
Beads Microcarriers
Concentrated high quality magnetite nanoparticles; provided with full COA. PVP surface for stability
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Superparamagnetic properties at room temperature
Magnetite nanoparticles (Fe3O4), 20 nm, 20 mg/ml, PVP surface for stability. These iron oxide particles are precisely engineered and unagglomerated, and exhibit superparamagnetic properties at ambient temperaturess. The size and superparamagnetic properties of these magnetite nanoparticles give them uses in a wide array of fields, such as catalysis, biosensors, ferrofluids, magnetic separation, and MRI contrast agents.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty