Would you like to log in again?
BioPure Platinum Nanoparticles 1 mg/ml Citrate (bare/displaceable surface), nanoComposix
Supplier: nanoComposix
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PTCB50-5M PTCB50-30M PTCB50-1M PTCB30-30M PTCB70-10M PTCB5-30M PTCB5-1M PTCB30-1M PTCB70-1M PTCB5-10M PTCB70-30M PTCB30-10M PTCB70-5M PTCB50-10M PTCB5-5M PTCB30-5M
103820-410EA 306.92 USD InStock
103820-410 103820-400 103820-422 103820-412 103820-424 103820-402 103820-414 103820-404 103820-426 103820-416 103820-428 103820-406 103820-430 103820-420 103820-418 103820-408
BioPure Platinum Nanoparticles 1 mg/ml Citrate (bare/displaceable surface), nanoComposix
Beads Microcarriers
Purified, concentrated, high quality platinum nps; sterile and endotoxin-free. Citrate surface.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials, extensively purified to remove residual reactants
  • Manufactured sterile
  • Provided in ultra to pure (Milli to Q) DI water
  • BioPure material is ideal for in to vitro and in to vivo experiments, nanotoxicology
Platinum nanoparticles, sizes from 5 to 70 nm, provided at 1 mg/ml (high concentration). PVP surface provides strong steric stability (40 kD PVP). BioPure materials are Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered, unagglomerated nanospheres with narrow size distributions (CV <20%).
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis.
Platinum nanoparticles have uses in many fields and applications, including catalysis, SERS, and nanotoxicology.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty