Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NanoXact Silica Nanospheres, Silica Surface, nanoComposix

Supplier: nanoComposix

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SISN120-100M SISN100-25M SISN50-100M SISN60-25M SISN20-500M SISN20-25M SISN500-500M SISN200-500M SISN80-25M SISN100-100M SISN100-1L SISN1000-1L SISN120-1L SISN200-25M SISN120-25M SISN60-500M SISN1000-100M SISN300-500M SISN200-100M SISN100-500M SISN80-1L SISN300-100M SISN80-500M SISN20-100M SISN50-500M SISN500-25M SISN60-100M SISN200-1L SISN500-100M SISN300-25M SISN60-1L SISN500-1L SISN120-500M SISN1000-500M SISN80-100M SISN20-1L SISN50-25M SISN1000-25M SISN300-1L SISN50-1L
103820-784EA 7667.69 USD InStock
103820-784 103820-786 103820-764 103820-766 103820-788 103820-800 103820-768 103820-802 103369-706 103369-726 103820-780 103820-782 103370-102 103369-686 103369-702 103820-804 103820-806 103820-808 103820-774 103820-796 103820-776 103820-798 103820-810 103820-778 103820-812 103370-098 103820-790 103820-792 103820-770 103820-772 103820-794 103369-710 103369-698 103369-730 103369-714 103820-814 103369-690 103820-816 103820-818 103369-694
NanoXact Silica Nanospheres, Silica Surface, nanoComposix
Beads Microcarriers
High quality silica nanoparticles, provided with full COA for great value. Silica (silanol) surface.

  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Silica surface offers robust and versatile surface for a wide variety of applications

Spherical silica nanoparticles are available in 20 to 400 nm sizes, in water

Each batch is extensively characterized to ensure it meets the highest quality standards, providing reliable and reproducible results. Products are supplied with COA's that include batch-specific TEM, DLS, Zeta Potential, UV-Vis, and pH measurements.

Silica nanoparticles’ large surface area can be used for Absorption Studies, Nanosphere Lithography, and Coating applications. The silanol surface can be modified using standard silane chemistry to introduce specific functional groups or to create hydrophobic or fluorophillic surface coatings.

Custom formulations and ISO13485/cGMP compliant materials can be provided on request.

Delivery information: Ships at ambient temperatures, via FedEx Priority Overnight (domestic).

Caution: DO NOT FREEZE. Store at 25°C, bath sonicate if needed. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty