Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NanoXact Silica-Shelled Silver Nanoplates 1 mg/ml Silica Shell (High Stability), nanoComposix

Supplier: nanoComposix

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SPSH800-30M SPSH980-5M SPSH1064-1M SPSH1064-10M SPSH980-10M SPSH980-1M SPSH1064-30M SPSH800-5M SPSH980-30M SPSH800-1M SPSH660-1M SPSH800-10M SPSH660-30M SPSH1064-5M SPSH660-10M SPSH660-5M
103820-894EA 306.92 USD InStock
103820-894 103820-884 103820-896 103820-886 103820-898 103820-910 103820-888 103820-900 103820-912 103820-890 103820-892 103820-902 103820-914 103820-904 103820-906 103820-908
NanoXact Silica-Shelled Silver Nanoplates 1 mg/ml Silica Shell (High Stability), nanoComposix
Beads Microcarriers
High concentration Ag nanoplates with peak absorbances from 550 to 1050 nm. Versatile silica surface

  • Silver nanoplates with peak wavelengths across 660 to 1064 nm
  • High quality nanoparticles, precisely engineered and highly characterized
  • Ultra strong light absorbing and scattering materials (OD's of ~100 to 160)
  • High concentration 1 mg/ml nanomaterial
  • Silica shell offers robust and versatile surface for a wide variety of applications

Platelet silver nanoparticles (a.k.a. silver nanoplates or silver nanodisks). Peak surface plasmon resonances (SPR) across wavelengths from 660 to 1064 nm. Provided at 1 mg/ml high concentration. Precisely engineered, unagglomerated nanoplates. Ultra to strong light absorbing and scattering materials.

Silica shell offers high stability in a variety of solvents and a versatile surface for attaching functional groups or creating hydrophobic & hydrophilic surfaces. Silica shell allows easier deposition onto substrates and incororation into many different solvents, resins, and curable materials because the silica shell provents metal nanoparticles from touching directly / aggregating, helping to preserve the particles' nano and optical properties.

Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis.

Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.

Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.

Caution: DO NOT FREEZE. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty