Would you like to log in again?
Human PSMA-coupled Magnetic Beads, Acro Biosystems
Supplier: Acro Biosystems
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MBPK0072MG MBPK00710MG
103871-290EA 1055.38 USD InStock
103871-290 103871-288
Human PSMA-coupled Magnetic Beads, Acro Biosystems
Beads Magnetic Beads
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The Human PSMA-coupled Magnetic Beads is produced by coupling biotinylated human PSMA protein to streptavidin-conjugated magnetic beads. The pre-coupled beads are ready to use for capturing anti-PSMA antibody from your sample with high specificity. The uniform size and large surface-to-volume ratio of the beads ensure highly efficent immunocapture in a simple, fast and convenient workflow with high reproducibilty of data.
Antibody Purification:
1. Resuspend the lyophilized beads by adding the buffer of choice.
2. Add analyte to the suspension, mix and incubate to enable specific binding of the beads and the target protein.
3. Magnetize beads, remove supernatant, and wash unbound protein fractions to capture target protein-bound beads.
4. Wash, magnetize the beads and collect purified target protein for use in downstream applications.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty