Would you like to log in again?
Human BCMA-coupled Magnetic Beads, Acro Biosystems
Supplier: Acro Biosystems
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MBSK00410MG MBSK0042MG
103871-344EA 861.54 USD InStock
103871-344 103871-342
Human BCMA-coupled Magnetic Beads, Acro Biosystems
Beads Magnetic Beads
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The Human BCMA-coupled Magnetic Beads is produced by coupling biotinylated Human BCMA protein to streptavidin-conjugated magnetic beads. The pre-coupled beads are ready to use for capturing anti-BCMA antibody from your sample with high specificity. The uniform size and large surface-to-volume ratio of the beads ensure highly efficent immunocapture in a simple, fast and convenient workflow with high reproducibilty of data.
Application Method:
a) Reconstitute the Beads following the COA. Wash and re-suspended the beads to a certain concentration by adding your dilution buffer.
b) Add the prepared beads to your samples.
c) Beads can be separated from your samples afterwards using a magnetic plate.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty