Would you like to log in again?
Human PPAR Panel Luciferase Reporter Assay Kit
Supplier: INDIGO Biosciences
IB00131-32P
103881-492KT 1593.85 USD InStock
103881-492
Human PPAR Panel Luciferase Reporter Assay Kit
Assays Cellular Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. Human PPAR Reporter Assays Panel reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert.
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
INDIGO's Human PPAR Panel (PPAR alpha, PPAR delta, and PPAR gamma) assay kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to reporter cells for each PPAR, optimized growth media, media for diluting test compounds, a control agonist for each PPAR, luciferase detection reagent, detailed protocol, and a protocol quick guide, and a cell culture-ready assay plate in strip-well format so that (if preferred) PPAR alpha, PPAR delta, and PPAR gamma assays may be performed at different times.
This panel contains sufficient materials to perform 32 PPAR alpha assays, 32 PPAR gamma assays, and 32 PPAR delta assays, all in a single 96-well plate format.
Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery information: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty