Would you like to log in again?
Human LXR Panel Luciferase Reporter Assay Kit
Supplier: INDIGO Biosciences
IB00321-48P
103881-510KT 1593.85 USD InStock
103881-510
Human LXR Panel Luciferase Reporter Assay Kit
Assays Cellular Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Human LXR Reporter Assays Panel All-inclusive Cell-Based Luciferase Reporter Assay Kit includes reporter cells for each LXR, optimized growth media, media for diluting test compounds, a control agonist for each LXR, luciferase detection reagent, a cell culture-ready assay plate in strip-well format.
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
INDIGO’s Human Liver X Receptor Panel (LXR alpha, LXR beta) assay kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to reporter cells for each LXR, optimized growth media, media for diluting test compounds, a control agonist for each LXR, luciferase detection reagent, a cell culture-ready assay plate in strip-well format so that (if preferred) LXR alpha and LXR beta assays may be performed at different times, detailed protocol, and a protocol quick guide.
This panel contains sufficient materials to perform 48 LXR alpha assays and 48 LXR beta assays, all in a single 96-well plate format.
Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery information: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty