Would you like to log in again?
Human RAR Panel Luciferase Reporter Assay Kit
Supplier: INDIGO Biosciences
IB02301-32P
103881-594KT 1593.85 USD InStock
103881-594
Human RAR Panel Luciferase Reporter Assay Kit
Assays Cellular Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Human RAR Reporter Assays Panel All-inclusive Cell-Based Luciferase Reporter Assay Kit, Including RARa, RARb, RARg, each in 1×32 96-well format.
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
INDIGO's Retinoic Acid Receptors Panel (RAR alpha, RAR beta, and RAR gamma) assay kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to reporter cells for each RAR, optimized cell culture medium, a medium for diluting test compounds, a control agonist for each RAR, stable-glow luciferase detection reagent, detailed protocol, Protocol Quick Guide, and a cell culture-ready assay plate in strip-well format so that (if preferred) RAR alpha, RAR beta, and RAR gamma assays may be performed at different times.
This panel contains sufficient materials to perform 32 RAR alpha assays, 32 RAR beta assays, and 32 RAR gamma assays in 96-well plate format.
Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery information: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty