Would you like to log in again?
In Vitro Screening for Drug-Induced Hepaotoxicity Assay Kit
Supplier: INDIGO Biosciences
ULC1003-48
103881-812KT 1132.31 USD InStock
103881-812
In Vitro Screening for Drug-Induced Hepaotoxicity Assay Kit
Assays Cellular Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from INDIGO’s Drug-Induced Hepaotoxcity Assay Using upcyte Hepatocytes All-inclusive Cell-Based Luciferase Reporter Assay Kit, 2×48 format.
  • Rapidly screen test compounds to identify those that induce acute liver cell toxicity
  • Kit includes two aliquots of cryopreserved Luminescent Reporter Hepatocytes (upcyte®), donor 10-13, isolated from an adult Caucasian female, that have been further modified to constitutively express the luciferase enzyme
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
The principal application of this assay is to rapidly screen test compounds to identify those that induce acute liver cell toxicity. This kit includes two aliquots of cryopreserved Luminescent Reporter Hepatocytes (upcyte®), donor 10 to 13, isolated from an adult Caucasian female, that have been further modified to constitutively express the luciferase enzyme.
In addition to two aliquots of the reporter hepatocytes, the kit provides two cell culture-ready assay plates, optimized Cell Culture Media (CCM) for use in all steps of the assay procedure (cell thawing, seeding, and preparation of the treatment media), luciferase detection reagent, and a reference compound that provides a positive control for hepatotoxicity.
The reagents and materials provided in this assay kit are formatted to allow the user to choose between two alternative assay setups. In one scenario 48 culture wells may be setup at two different times. In the other assay scenario 96 culture wells may be setup at one time.
Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery information: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty