Would you like to log in again?
Anti-SARS-CoV-2 (P.1) Neutralizing Antibody Titer Serologic Assay Kit (Spike RBD), Acro Biosystems
Supplier: Acro Biosystems
RASN03496TESTS
103888-182EA 700 USD InStock
103888-182
Anti-SARS-CoV-2 (P.1) Neutralizing Antibody Titer Serologic Assay Kit (Spike RBD), Acro Biosystems
Assays Immunological Assays
This kit is developed for qualitative detection or titer measurement of Anti-SARS-CoV-2 (P.1) neutralizing antibody (Spike RBD) in human serum.
Multiple variants of SARS-CoV-2 are circulating globally and posting new challenges to human health. As concerns over the potential impacts of the variants grow, people ask if one possesses protective neutralizing antibodies (NAbs) against the variants after receiving the currently available vaccines; and if the NAb therapeutic agents are equally effective to treat the mutant infection as the wild type. To facilitate the mutant-related research, drug trials and vaccine assessments, a high-throughput assay to measure neutralizing antibodies against the mutants is in urgent need.
Your experiment will include five simple steps:
a) All reagents were returned to room temperature (20 to 25 °C) before use.
b) Make series the tested sample and control with ditlution buffer, HRP-SARS-CoV-2 Spike RBD diluted with dilution buffer.
c) Add the diluted sample, Control and the HRP-SARS-CoV-2 Spike RBD add the plate respectively.
d) Wash the plate and add TMB or other colorimetric HRP substrate.
e) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculated as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty