Would you like to log in again?
Anti-SARS-CoV-2 (B.1.1.7) Antibody IgG Titer Serologic Assay kit (Spike RBD), Acro Biosystems
Supplier: Acro Biosystems
RAST02696TESTS
103888-198EA 700 USD InStock
103888-198
Anti-SARS-CoV-2 (B.1.1.7) Antibody IgG Titer Serologic Assay kit (Spike RBD), Acro Biosystems
Assays Immunological Assays
This kit is developed for serologic test for IgG titer of Anti-SARS-CoV-2 Spike RBD antibody in serum/plasma in vitro. The Spike RBD antigen used in this kit contains the N501Y mutation found in the U.K. variant (known as B.1.1.7, 20B/501Y.V1 or VOC 202012/01), the South African variant (known as B.1.351 or 20C/501Y.V2) and the Brazilian variant (known as P.1).
Multiple variants of SARS-CoV-2 are circulating globally and posting new challenges to human health. The variant B.1.1.7 identified in the United Kingdom (UK) carries a large number of mutations and is reported to spread more easily and quickly than other variants. The variant B.1.351 identified in South Africa and P.1 identified in Brazil show evidence of increased transmissibility and resistance to established immunity. A common mutation identified in the U.K., South African and Brazilian variants is N501Y, which resides on the receptor-binding domain (RBD) of SARS-CoV-2 spike protein. To evaluate the impacts of the new variants, a rapid and effective assay kit detecting the levels of antibody against the mutant is in urgent need.
Your experiment will include four simple steps:
a) Add your sample to the plate. The samples and Control sample are diluted by Dilution Buffer.
b) Add diluted Secondary antibody HRP-Anti-Human IgG to the plate. The Secondary antibody is diluted by Dilution Buffer.
c) Wash the plate and add TMB or other colorimetric HRP substrate.
d) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculated as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty