Would you like to log in again?
Mouse Anti-SARS-CoV-2 Antibody IgG Titer Serologic Assay kit, Acro Biosystems
Supplier: Acro Biosystems
RAST01896TESTS
103888-718EA 624.62 USD InStock
103888-718
Mouse Anti-SARS-CoV-2 Antibody IgG Titer Serologic Assay kit, Acro Biosystems
Assays Immunological Assays
This kit is developed for serologic test for Mouse IgG titer of Anti-SARS-CoV-2 antibody in serum/plasma in vitro.
The ongoing SARS-COV-2 pandemics has sickened 3.7 million people around the world, claiming over 1000000 lives. To deal with this global public health crisis, unprecedented efforts are being made to study the virus, SARS-CoV-2. At present, there are 193 new crown vaccine projects registered with the World Health Organization are being developed. To support these studies, ACRO has developed Anti-SARS-CoV-2 subtype antibody Serologic Assay kit including:IgG1, IgG2 IgG3, IgG4.
Your experiment will include four simple steps:
a) Add your sample to the plate, take theAnti-SARS-CoV-2 Antibody (Control, Mouse IgG )as Control sample. The samples and Control sample are diluted by Dilution Buffer.
b) Add diluted Secondary antibody HRP-Goat anti-Mouse IgG to the plate. The Secondary antibody is diluted by Dilution Buffer.
c) Wash the plate and add TMB or other colorimetric HRP substrate.
d) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculated as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty