Would you like to log in again?
SARS-CoV-2 Spike RBD Titer Assay Kit, Acro Biosystems
Supplier: Acro Biosystems
RASA02196TESTS
103888-866EA 624.62 USD InStock
103888-866
SARS-CoV-2 Spike RBD Titer Assay Kit, Acro Biosystems
Assays Protein Assays
This kit is developed for detecting SARS-CoV-2 Spike RBD in the sample.
The newly identified Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) has posed a serious threat to human health. A rapid and effective assay kit detecting the levels of SARS-CoV-2 Spike Protein RBD is urgently needed to accelerate the development of COVID-19 vaccines.
Your experiment will include six simple steps:
a)All reagents were returned to room temperature(20 to 25 °C) before use.
b) Add your sample to the plate, take the SARS-CoV-2 Spike RBD as Control sample. The samples and Control sample are diluted by Dilution Buffer.
c) Add a diluted Secondary antibody biotin-Anti-SARS-CoV-2 Spike RBD Antibody to the plate. The Secondary antibody is diluted by Dilution Buffer.
d) Add a diluted Streptavidin-HRP to the plate.
e) Wash the plate and add TMB or other colorimetric HRP substrate.
f) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculated as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of protein bound.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty