Would you like to log in again?
SARS-CoV-2 Spike Trimer (D614G)-coupled Magnetic Beads, Acro Biosystems
Supplier: Acro Biosystems
MBSK0162MG MBSK01610MG
103889-378EA 1400 USD InStock
103889-378 103889-376
SARS-CoV-2 Spike Trimer (D614G)-coupled Magnetic Beads, Acro Biosystems
Beads Magnetic Beads
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The pre-coupled magnetic beads coupled with biotinylated SARS-CoV-2 Spike trimer(D614G) protein to streptavidin conjugated magnetic beads, which can capture the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample. The beads are in uniform size, narrow size distribution with large surface area and unique surface coating, which can help you get the best performance and highly reproducible results. This very first SARS-CoV-2 Spike trimer(D614G) protein-coupled magnetic beads will bring great convenience with minimum non-specific binding and developed protocols. This ready to use products could greatly save your time and hassle.
Application Method:
a) Reconstitute the Beads following the CoA, wash the Beads and suspended to a certain concentration by adding assay Buffer.
b) Add your antibody or ACE-2 protein to the beads, incubation and wash the beads.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty