Would you like to log in again?
SARS-CoV-2 Nucleocapsid Protein Titer Assay Kit, Acro Biosystems
Supplier: Acro Biosystems
RASA01096TESTS
103889-730EA 624.62 USD InStock
103889-730
SARS-CoV-2 Nucleocapsid Protein Titer Assay Kit, Acro Biosystems
Assays Protein Assays
This kit is developed for detecting the SARS-CoV-2 Nucleocapsid Protein in cell supernatant or SARS-CoV-2 virus in vitro.
Nucleocapsid (N) protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein important for viral genome replication and modulation of cell signaling pathways. It was first identified by a research team while they were screening for ADP-ribosylated proteins during coronavirus (CoV) infection (Grunewald M. E., et al. 2017, Virology; 517: 62-68). The array of diverse functional activities accommodated in N protein makes it more than a structural protein but also an interesting target in the development of antiviral therapeutics. Because of the conservation of N protein sequence and its strong immunogenicity, N protein of coronavirus is chosen as a diagnostic tool.
Your experiment will include 5 simple steps:
a) Add your sample to the plate, take the SARS-CoV-2 Nucleocapsid protein as Control sample. The samples and Control sample are diluted by Lysis Buffer.
b) Add a diluted Secondary antibody biotin-anti-SARS-CoV-2 Nucleocapsid Antibody to the plate. The Secondary antody is diluted by Dilution Buffer.
c) Add a diluted Streptavidin-HRP to the plate.
d) Wash the plate and add TMB or other colorimetric HRP substrate.
e) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculate as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty