Would you like to log in again?
Anti-SARS-CoV-2 Total Antibody Titer Serologic Assay kit (Spike RBD), Acro Biosystems
Supplier: Acro Biosystems
RAST00996TESTS
103889-732EA 624.62 USD InStock
103889-732
Anti-SARS-CoV-2 Total Antibody Titer Serologic Assay kit (Spike RBD), Acro Biosystems
Assays Immunological Assays
This kit is developed for serologic test for Total Anti-SARS-CoV-2 antibody in serum/plasma in vitro.
The newly identified Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) has posed a serious threat to human health. A rapid and effective assay kit detecting the levels of anti-SARS-CoV-2 in human serum can facilitate research on characterization of antibodies produced in response to SARS-CoV-2 infection.
Your experiment will include 4 simple steps:
a) Add your sample to the plate. The samples and Control sample are diluted by Dilution Buffer.
b) Add diluted HRP-SARS-CoV-2 Spike RBD to the plate. The HRP-SARS-CoV-2 Spike RBD is diluted by Dilution Buffer.
c) Wash the plate and add TMB or other colorimetric HRP substrate.
d) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculate as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.
Certifications: ISO 9001 and ISO 13485.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty