Would you like to log in again?
Blue Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Curad
Sepharose™
90100123
104013-192PK 2480.82 USD InStock
104013-192
Blue Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
This media is suitable for affinity chromatography.
Chemical stability: Commonly used buffers, 70% ethanol, 8 M urea and 6 M guanidine hydrochloride.
Storage solution: 20% ethanol
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty