Would you like to log in again?
Endonuclease IV (NFO) 5,000 Units, Intact Genomics
Supplier: Intact Genomics
3425
104013-272EA 340.38 USD InStock
104013-272
Endonuclease IV (NFO) 5,000 Units, Intact Genomics
Enzymes
Endonuclease IV (Nfo) from Escherichia coli is a 32 kD metalloprotein that aids in the repair of damaged DNA. The enzyme functions both as an apurinic/apyrimidinic nuclease (1) and as a 3′-terminal di-esterase (1-4).
  • No detectable exonuclease or DNA N-glycosylase activity
  • Useful for single cell gel electrophoresis, alkaline elution, and alkaline unwinding
Its 3′-terminal di-esterase activity is important in the repair of DNA strand breaks generated by oxidation and ionic radia-tion (2, 3). In such events, the strand breaks terminate with either a 3′ phosphate or a deoxyribose fragment, preventing repair by DNA polymerase I or DNA ligase. Endonuclease IV removes the blocking groups, leaving a free 3′-hydroxyl terminus. This enzyme does not have detectable associated exonuclease or DNA N-glycosylase activity (1).
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty