Would you like to log in again?
JM109(De3) Chemically Competent Cells, Intact Genomics
Supplier: Intact Genomics
1063-48 1063-24
104013-282PK 294.23 USD InStock
104013-282 104013-280
JM109(De3) Chemically Competent Cells, Intact Genomics
Competent Cells
Intact Genomics JM109(DE3) chemically competent Escherichia coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. JM109(DE3) is useful for the high-level expression of genes cloned into vectors for expression of sequence downstream from the T7 promoter.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty