Would you like to log in again?
Lba4404 Chemically Competent Agrobacterium, Intact Genomics
Supplier: Intact Genomics
1085-06 1085-18
104013-294PK 605.77 USD InStock
104013-294 104013-292
Lba4404 Chemically Competent Agrobacterium, Intact Genomics
Competent Cells
Intact Genomics LBA4404 Chemically Competent Agrobacterium cells are useful for transgenic operations of tomatoes, tobacco and other plants.
  • High transformation efficiencies
  • Especially useful for cDNA gDNA library construction
Intact Genomics LBA4404 chemically competent Agrobacterium cells are optimized for the highest transformation efficiencies which is ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction. The LBA4404 strain is useful for transgenic operations of tomatoes, tobacco and other plants. LBA4404 contains a rifampicin resistance gene (rif). LBA4404 strain also contains a octoprine-type Ti plasmid pAL4404 without self-transport function, which contains the vir gene.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty