Would you like to log in again?
Eha105 Electrocompetent Agrobacterium, Intact Genomics
Supplier: Intact Genomics
1284-36 1284-12
104013-312PK 633.46 USD InStock
104013-312 104013-310
Eha105 Electrocompetent Agrobacterium, Intact Genomics
Competent Cells
Intact Genomics EHA105 electrocompetent Agrobacterium cells are useful for transgenic operations of rice, tobacco and other plants.
  • High transformation efficiencies
  • Especially useful for cDNA gDNA library construction
Intact Genomics EHA105 electrocompetent Agrobacterium cells are optimized for the highest transformation efficiencies which is ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction. The EHA105 strain is useful for transgenic operations of rice, tobacco and other plants. EHA105 contains a rifampicin resistance gene (rif). EHA105 strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function, which contains the vir gene.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty