Would you like to log in again?
Ig-Fusion Cloning Enzyme Premix, Intact Genomics
Supplier: Intact Genomics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4117-1 4115-1 4111-1
104013-324EA 865.38 USD InStock
104013-324 104013-320 104013-322
Ig-Fusion Cloning Enzyme Premix, Intact Genomics
Nucleic Acid Reagents Cloning Kits and Ligation
Intact Genomics propriety Ig-Fusion™ cloning technology is a simple, rapid and highly efficient cloning kit which allows to directly clone any PCR product(s) to any linearized expression vector at any site.
The PCR fragments can be generated by Intact Genomics high fidelity Pfu DNA polymerase or other high fidelity DNA polymerases, with primers having 15 to 18 bases of homology at their linear ends to where the product need to fuse. The linearized vector can be generated by PCR or restriction enzymes. The kit is so robust that multiple DNA fragments can be assembled simultaneously and cloned into one construct in a single reaction step within short times (usually 10-30 min) with more than 95% cloning efficiency.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty