Would you like to log in again?
DirectPlate™ BL21(DE3) Chemically Competent Escherichia coli Cells, Intact Genomics
Supplier: Intact Genomics
1019-12 1019-36
104013-346PK 190.38 USD InStock
104013-346 104013-348
DirectPlate™ BL21(DE3) Chemically Competent Escherichia coli Cells, Intact Genomics
Competent Cells
DirectPlate™ BL21(DE3) chemically competent Escherichia coli cells are a perfect choice for researchers looking to simplify their transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate!
  • No heat shock
  • Save time (3 minutes transformation instead of a 90 minute transformation)
  • High transformation efficiency
DirectPlate™ BL21(DE3) chemically competent Escherichia coli cells provide higher transformation efficiency than any competitor‘s similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning, protein expression and sub-cloning. Intact Genomics (ig®) DirectPlate™ competent cells offer simple, fast, and robust results for your DNA transformation needs. Genotype: F–ompT hsdS(rB– mB–) gal dcm λ(DE3).
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty