Would you like to log in again?
S-Chip Disposable Sperm Counting Slides
Supplier: Bulldog Bio
S-Chip
DCS-S01
104016-462PK 423.08 USD InStock
104016-462
S-Chip Disposable Sperm Counting Slides
Slides Chamber Slides
When it comes to in vitro fertilization (IVF) and intrauterine insemination (IUI), accurate sperm counts are vital. The S-Chip for semen testing provides exact and reliable counting while simplifying the process.
  • All-in-one disposable hemocytometer for counting sperm
  • Easy and precise counting
  • Individually sealed packaging
  • Lot tested and certified
The S-Chip contains two quartz grade optically engineered 20 µm height counting chambers that do not need coverslips. Simple operation involves pipetting the sample into each chamber, placing on the microscope, and then counting. Dispose once finished.
Packaging: 100 tests per box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty