Would you like to log in again?
Powerblanket® Lite Gas Cylinder Heaters (Propane)
Supplier: POWERBLANKET LLC MS
Powerblanket® Lite
PBL100 PBL500 PBL1K PBL420 PBL20
104018-918EA 913.61 USD InStock
104018-918 104018-928 104018-930 104018-900 104018-910
Powerblanket® Lite Gas Cylinder Heaters (Propane)
Heaters Heating Mantles
Powerblanket tank propane tank heaters optimize container pressure and increase tank efficiency. Powerblanket products provide a maintenance-free heating solution, which eliminates the headache of maintaining critical temperatures. By using Powerblanket propane tank heaters, gas cylinders will last longer and be more effective. This saves time, hassle, and money.
  • Quickly heats cylinder to a preset 90 °F / 32 °C (±10 °F/5 °C)
  • Provide an insulated full-wrap design
  • Increase flow rate performance and efficiency of gas cylinders
  • Saves money by optimizing gas and material usage<
  • Eliminate unnecessary cylinder refills in cold weather
  • Powerblanket Lite - designed for light residential and industrial applications, low power density
  • Powerblanket - designed for any definitive heating applications, surrounded by a rugged vinyl shell, safe to use in temperatures as low as −20 °F
  • Powerblanket Xtreme - designed for harsh climates, safe to use in temperatures as low as −40 °F
Cold weather can wreak havoc on propane tanks and other gas cylinders. When the temperature drops, it’s very difficult to maintain optimal pressure in a gas cylinder. This prevents the propane from vaporizing which in turn makes your propane tank unusable. Powerblanket offers a solution with propane tank heaters for controlling temperatures of propane, as well as butane, nitrogen, oxygen and other compressed gases. These heaters provide a uniform barrier of heat across the entire cylinder with the help of a fixed internal thermostat.
Certifications: Certified to by ETL to UL and CSA safety standards.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty