Would you like to log in again?
Powerblanket® Heater Blankets for Gas Cylinders
Supplier: POWERBLANKET LLC MS
Powerblanket®
GCW420 GCW20 GCW30 GCW100 GCW40
104018-938EA 697.85 USD InStock
104018-938 104018-904 104018-922 104018-998 104018-964
Powerblanket® Heater Blankets for Gas Cylinders
Heaters Heating Mantles
These blankets help to prevent the gas from stratifying, this in turn makes the gas standard stable.
  • Increase performance and efficiency of gas cylinders
  • Insulated full-wrap design
  • Reduce downtime and increase profitabablity
  • Uniform distribution of heat
  • Highly efficient, low energy consumptions
  • Class 1 Division 2 blankets
Powerblanket® offers a variety of gas cylinder heaters for controlling temperatures of propane, butane, nitrogen, oxygen and other compressed gases.
Powerblanket® gas cylinder heaters provide a uniform barrier of heat across the entire cylinder. The superior heating solution reduces costs by optimizing container temperatures and increasing cylinder efficiency.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty