Would you like to log in again?
CO₂ Shaking Incubator, Benchtop
Supplier: Alkali Scientific
RS8095
104026-760EA 12583.06 USD InStock
104026-760
CO₂ Shaking Incubator, Benchtop
Shakers and Mixers Shaking Incubators
Benchtop CO₂ shaking incubator is designed to maintain a stable environment for your cell cultures and to minimize contamination. With our unique and proprietary heat distribution technology, the ST series of CO₂ incubator provides the highest levels of temperature accuracy with minimal fluctuation.
  • Precision temperature control, six-sided heating
  • Precise monitoring over CO₂ with dual beam IR sensor
  • Seamless, stainless-steel chamber
  • Digital control of humidity percentage (PLUS models only)
The six-side heating system, in combination with a low speed internal fan, uniformly spreads air throughout the chamber (without air exchange from the external environment). The chamber walls are constructed of high grade (316) stainless-steel, and all corners are rounded, preventing the accumulation of dirt/debris. For added security, model feature an internal alarm to alert the user if the CO₂ percentage changes, or the temperature fluctuates by more than 1 °C (33.8 °F) from the set point.
In addition, a 'quick-cal' calibration button allows for quickly adjusting the chamber temperature to match that of a properly calibrated external thermometer. PLUS models include advanced programming for high heat decontamination (at 125 °C (257 °F)), a unique split inner glass door for access to the chamber without exposing it fully to the external environment and programmable humidity percentage, up to 95%.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty