Would you like to log in again?
CellPro™ Neutralizing Solution, Alkali Scientific
Supplier: ALKALI SCIENTIFIC MS
DS0215-4L DB0319-20L DS0215 DB0319
104027-500EA 72.29 USD InStock
104027-500 104027-480 104028-868 104026-840
CellPro™ Neutralizing Solution, Alkali Scientific
Buffers Cell Culture Buffers
CellPro™ Neutralizing Solution is an essential reagent in molecular biology and biochemical experiments, particularly in protocols involving nucleic acid hybridization and blotting techniques.
  • Sterile
  • Filtered
This solution is specifically formulated to neutralize the effects of strong bases or acids used during the denaturation process. For example, after treating samples with denaturing solutions such as 0.5 M NaOH with 1.5 M NaCl, the CellPro™ Neutralizing Solution effectively restores the pH balance, creating an optimal environment for subsequent experimental steps.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty