Would you like to log in again?
CellPro™ Calcium Chloride, 1M, Alkali Scientific
Supplier: ALKALI SCIENTIFIC MS
DB0238
104027-392EA 50.75 USD InStock
104027-392
CellPro™ Calcium Chloride, 1M, Alkali Scientific
Buffers Cell Culture Buffers
Alkali Scientific™ CellPro™ Calcium Chloride, 1M, is a meticulously prepared solution designed to meet the demands of various molecular biology and biochemical applications.
  • Triple-distilled
  • Sterile
With its sterile and filtered formulation, this solution ensures exceptional purity and reliability, essential for precise laboratory experiments. Calcium chloride is a versatile reagent widely used in molecular biology laboratories for tasks such as DNA transformation, protein crystallization, and enzyme activation. The meticulously controlled formulation and stringent quality control measures of CellPro™ Calcium Chloride guarantee consistent and reproducible results, enabling researchers to pursue their scientific inquiries with confidence. Trust in the purity and reliability of CellPro™ Calcium Chloride for your laboratory's molecular biology and biochemical needs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty