Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NDEL1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-5524R
10417-114EA 649.67 USD InStock
10417-114
Anti-NDEL1 Rabbit Polyclonal Antibody
Antibodies
Nudel is important for normal cortical development. It is invovled in microtubule organization, nuclear translocation, and neuronal positioning in concert with various other factors (including Lis1, Pafah1b1, Pahfah1b2, dynein, dynorphin A and cdk5). Western blot analysis of mouse tissues shows abundant expression of Nudel in brain and testis, and much lower expression in heart, liver, kidney, and skeletal muscle. In fractionated rat brain, Nudel and Lis1 are both found in fractions enriched for postsynaptic density proteins. Immunostaining of embryonic day 18 mouse brain sections revealed staining of migrating neurons and thalamocortical axons of the intermediate zone of the developing cerebral cortex, as well as several other developing brain regions. The deduced protein contains 345 amino acids and has a calculated molecular mass of 38.4 kDa. It has a coiled coil motif (residues 19 to 201), followed by several potential phosphorylation sites for casein kinase II, protein kinase C or CDK5. Nudel shares about 50% identity with mouse and human NUDE proteins.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NDEL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty