Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP28 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-5876R
10425-638EA 621.95 USD InStock
10425-638
Anti-MMP28 Rabbit Polyclonal Antibody
Antibodies
Matrix Metalloproteinase 28, also known as epilysin, was first cloned from human skin keratinocytes and described to reflect its role in the remodeling of the epidermis. It was found in testis, as well as heart, brain, placenta, lung, prostate, intestine, and colon. MMP28 was later cloned from human lung and found in lung, kidney, brain, skeletal muscle, and several tumor cell lines. At least three MMP28 transcripts of 2.6, 2.0, and 1.2 kb have been reported possibly representing alternatively spliced forms of MMP28. There are two human sequences (isoform1 and isoform 2) which encode proteins of 520 and 393 amino acids with predicted masses of 58.9 and 44.5 kDa respectively. The mouse sequence contains two inserts of 41 and 39 amino acids respectively, relative to the human sequence. Mouse epilysin has a predicted molecular mass of approx. 70 kDa. Mouse and human epilysin are highly conserved and share 97% identical residues. Epilysin (MMP-28) contains the key domains of the other MMPs: a signal peptide, conserved cysteine-containing prodomain (with a furin cleavage site), conserved histidine-containing catalytic domain, hinge domain, and hemopexin domain. MMP28 has a furin cleavage site, similar to MMP11, and is cleaved by the prohormone convertase family of enzymes. Sequence identity between MMP28 and the other MMPs is low overall. MMP28 is most closely related to MMP19 (39% sequence identity). Recombinant MMP28 degrades casein.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMP28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty