Would you like to log in again?
Fluid Thioglycollate Medium in ReadyRack™, Disposable Tube Rack, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
K212BX K212BX
10436-340PK 580.43 USD InStock
10436-340 CA10436-340
Fluid Thioglycollate Medium in ReadyRack™, Disposable Tube Rack, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Fluid Thioglycollate Medium in ReadyRack™ is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms in normally sterile materials
  • 80 Tube Ready Rack
  • Holes in the bottom of the rack allow for sterilant to drain
  • Convenient hand-holds
  • No cardboard for low particle counts
The ReadyRack is a plastic rack designed for cleanroom use and can be rinsed with alcohol. It is a convenient way to bring media into a controlled area while avoiding outside contamination.
Simply remove ReadyRack from box outside of cleanroom. Take into cleanroom and remove plastic bag. Spray down entire ReadyRack with isopropyl alcohol, and allow to dry.
Ordering information: Fluid Thioglycollate Medium in ReadyRack, USP, contains 16 x 125 mm tubes, 10 mL fill. This product must be ordered by the package of 80.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty