Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STK17A Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-6770R-CY5.5
10454-346EA 792.04 USD InStock
10454-346
Anti-STK17A Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
DRAK1 (DAP kinase-related apoptosis-inducing protein kinase 1) is a novel member of the ser/thr protein kinase family, which mediate apoptosis through their catalytic activities. The full-length cDNA encodes a deduced 414-amino acid protein with a molecular mass of 46.56 kD. DRAKs contain an N-terminal kinase domain and a C-terminal regulation domain. DRAK1 messenger RNA appears to be ubiquitously expressed in human tissues. Overexpression of DRAK1 induces apoptosis. It has been shown in vitro that DRAK1 is capable of autophosphorylation and of phosphorylating the myosin light chain as an exogenous substrate, and that the noncatalytic C terminus is crucial for full kinase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STK17A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty