Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Marathon MicroDL™ Temperature Data Loggers, Therapak®
Supplier: Therapak
36574 36569 36573
10497-132EA 413.01 USD InStock
10497-132 10497-128 10497-134
Marathon MicroDL™ Temperature Data Loggers, Therapak®
Data Loggers
The Marathon MicroDL™ is a programmable data logger engineered to display realtime temperature that also offers the ability to scroll through a summary of recorded history information
The operating temperature range is –30 to 72 °C (–22 to 162 °F). MicroDL™ Reader and Software allows the data logger to be programmed and provides easy access to temperature graphs and data.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty