Would you like to log in again?
Gastroenterology (GI) Biopsy Collection and Transport Kits, Therapak®
Supplier: Therapak
38873
10497-202CS 243.5 USD InStock
10497-202
Gastroenterology (GI) Biopsy Collection and Transport Kits, Therapak®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
Both kits feature standardized components to maximize the ease of collecting and packing specimens for transport
Corrugated inserts are printed with the GI biopsy location ID for easy recognition. In addition, a set of jar labels with the location ID is included in the kit to ensure proper placement of the specimen jars at the point of packing and identification once received at the test location. Customization of these kits is available upon request.
Primary receptacles in the form of 20mL jars are 95kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation. The kits come with either two or six 20mL jars pre-filled with 10mL of formalin solution. Jars are secured by a corrugated insert and placed into the provided zip-style biohazard bag with absorbent material for transport.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty