Would you like to log in again?
Histology Bucket with Lid, Therapak®
Supplier: Therapak
75100G 75098G 75099G
10497-264CS 165.28 USD InStock
10497-264 10497-268 10497-266
Histology Bucket with Lid, Therapak®
Buckets
These polypropylene histology buckets are conveniently packed in case quantities and each bucket features a carrying handle and snap-on lid
The lid secures tightly to the bucket to reduce odor. The buckets are autoclavable, easy to stack, and are resistant to cracking when exposed to common cytology and histology based chemicals such as formaldehyde.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty