Would you like to log in again?
Inlet Filter Assembly Kit, Cytiva
Supplier: Cytiva
18-1113-15 18-1113-15
10497-990EA 235.18 USD InStock
10497-990 CA10497-990
Inlet Filter Assembly Kit, Cytiva
Filters Liquid Process Filters
Kit contents include one support net, one filter (mounted on fliterholder), and one screw lid and cap membrane.
The wetted parts, except for the membrane, are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, with the exception of 100% Ethyl acetate, 100% Hexane, and 100% Tetrahydrofuran.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty