Would you like to log in again?
Superloops, 10 and 50 ml, Cytiva
Supplier: Cytiva
18-1113-81 18-1113-82 18-1113-82 18-1113-81
10497-996EA 1730.19 USD InStock
CA10497-996 CA10497-994 10497-996 10497-994
Superloops, 10 and 50 ml, Cytiva
Chromatography Supplies Chromatography Spare Parts
Superloop can be used in conjunction with 7-port valves
  • Flexible: inject large or small sample volumes
  • High reproducibility and recovery
  • No gradient dilution with internal movable seal
The patented devices enable controlled loading of large samples, either in the form of multiple small injections or a single large injection. They may also be used to inject pulses of a specific buffer solution. For use with aqueous solutions and alcohols.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty