Would you like to log in again?
Sample Loop with Fingertight Connectors, Cytiva
Supplier: Cytiva
18-1114-02 18-1114-02 18-1114-01 18-1140-53 18-1140-53 18-1114-01
10498-020EA 291.94 USD InStock
10498-020 10498-000 CA10498-020 CA10497-998 10497-998 CA10498-000
Sample Loop with Fingertight Connectors, Cytiva
Chromatography Supplies Chromatography Sample Loops
The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100% Ethyl acetate, 100% Hexane, and 100% Tetrahydrofuran.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty