Would you like to log in again?
Affinity Chromatography Columns, Tagged Package, His, Cytiva
Supplier: Cytiva
29-0588-03 29-0588-03
10498-068EA 206.37 USD InStock
10498-068 CA10498-068
Affinity Chromatography Columns, Tagged Package, His, Cytiva
Chromatography Columns
His package is designed for the convenient purification of histidine-tagged proteins.
The His package comes with ready-to-use columns packed with high performance media developed for high yield (HisTrap™ HP, 1×1 ml) or high purity (HiTrap™ TALON crude, 1×1 ml) and a HiTrap™ desalting column (1×5 ml) for removing imidazole from eluate.
Delivery information: The kit contains one of each HisTrap™ HP (1×1 ml), HiTrap™ TALON crude (1×1 ml) and HiTrap™ Desalting (1×5 ml) columns.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty