Would you like to log in again?
Anti-CBP Tag Mouse Monoclonal Antibody [clone: 1D11E5]
Supplier: Genscript
GenScript®
A01798-40
10498-180EA 137.79 USD InStock
10498-180
Anti-CBP Tag Mouse Monoclonal Antibody [clone: 1D11E5]
Antibodies
GenScript CBP tag antibody, mAb, mouse recognizes CBP-tagged proteins.
  • Comprehensive pool
  • against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty