Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Phosphotyrosine Mouse Antibody

Supplier: Genscript
GenScript®
A01819-100
10498-210EA 396.34 USD InStock
10498-210
Anti-Phosphotyrosine Mouse Antibody
Antibodies
The antibody is specific for proteins containing phosphotyrosine residues and phosphotyrosine peptides. It does not cross-react with phosphoserine, phosphothreonine or nonphosphorylated tyrosine residues.

  • High specificit
  • High affinity
  • High stability

GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including sandwich ELISA and western blot.

Type: Primary
Antigen: Phosphotyrosine
Clonality:
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa, IgG2a kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty