Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
ELIMINASE®, Decontaminants, Decon Labs
Supplier: Decon Labs
1102 1101
10798-804CS 505.84 USD InStock
10798-804 10498-216
ELIMINASE®, Decontaminants, Decon Labs
Cleaning Agents Decontaminants
ELIMINase removes RNase, DNase, unwanted DNA and radioactive contamination.
  • Phosphate free and non-toxic
  • Safely disposable and totally rinsable for the decontaminated surface
  • Eliminates the time-consuming method of autoclaving
It works instantly so there is no waiting. ELIMINase is an ideal substitute for using DiEthylPryoCarbonate (DEPC) which is a carcinogen. ELIMINase can be used on all surfaces including glass, plastic, metal and ceramic.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty