Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BioBall® MultiShot, For Quantitative Quality Control Microorganisms, bioMerieux
Supplier: bioMérieux
BioBall®
412647 412540 412646 412541 56150 410386
97067-334PK 1412.62 USD InStock
97067-334 97067-332 10500-298 10500-296 97067-331 75996-728
BioBall® MultiShot, For Quantitative Quality Control Microorganisms, bioMerieux
Control reagents
BioBall® mixed kits contain the most relevant pharmacopoeia strains required for quantitative microbiology
BioBall® contains a precise number of microorganisms in a water soluble ball, delivering unprecedented accuracy for Quantitative Microbiological Quality Control. These BioBall® Multishot 550 are mean of between 500 and 600 cfu with a standard deviation of ≤ 10% of the mean. Designed for re-hydration into 1.1 mL of MultiShot 550 Re-Hydration Fluid to provide 10 x 100 µL doses of 50 cfu. Ideal for pharmaceutical microbiology QC where multiple positive controls in the 10 to 100 cfu range are needed.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty