Would you like to log in again?
Etest® Inoculator Retro C80, bioMerieux
Supplier: bioMérieux
559803
10500-472EA 1664.17 USD InStock
10500-472
Etest® Inoculator Retro C80, bioMerieux
Spreaders Inoculating Loops and Needles
Retro C80 is a rota-plater that simplifies and standardizes the inoculation of small and large agar plates making Etest easier to read when compared to manual streaking
  • Reduces operator fatigue
  • Makes readings easier
  • Standardizes inoculation
Allows easier application of samples to testing equipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty