Would you like to log in again?
ActivArmr® 97-011 High-Visibility Nitrile Gloves, 3/4 Dipped, Ansell
Supplier: Ansell
ActivArmr®
256064 112734 256067 256066
10510-828CS 430.99 USD InStock
10510-828 10510-826 10510-832 10510-830
ActivArmr® 97-011 High-Visibility Nitrile Gloves, 3/4 Dipped, Ansell
Gloves
Protecting workers from the elements, ActivArmr® 97-011 gloves deliver incomparable high-visibility hand protection for cold, wet environments.
  • Superior cold protection
  • Excellent handling of wet, dry, and oily materials
  • High-visibility gloves help avoid injuries
Colder temperatures impair reflex time, dexterity, grip and feel.
Visibility is a top priority in many work environments. It can help avoid injuries and save lives. ActivArmr® 97-011 Cold Weather Hi-Viz gloves are specially designed with an exclusive black nitrile coating that contrasts with a high-visibility orange liner in low-light and low-visibility conditions. The ActivArmr® 97-011 gloves protect the entire hand from the effects of wind, liquid and cold. The 97-011 is tested and proven to perform at ‒4 °F/‒20 °C. This best-in-class combination does it all without compromising the dexterity and movement that is essential for getting the job done.
The ActivArmr® 97-011 gloves combine superior protection from the elements, while the ¾ double-dipped nitrile coating has a sandy finish for excellent wet, oily and dry materials handling, preventing injury from slippage.
Certifications: ANSI Cut Level 2. ANSI Abrasion Level 4.
Packaging: Gloves are packaged 12 inners in a carton; 6 pairs in an inner carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty