Would you like to log in again?
VWR® Flat-Bottom Boiling Flasks
Supplier: VWR
058.202.60 058.202.59A
10536-638CS 483.07 USD InStock
10536-638 10536-648
VWR® Flat-Bottom Boiling Flasks
Flasks Flat Bottom Flasks
The flasks come with uniform wall thickness which makes these flasks ideal for heating.
  • Borosilicate glass fabrication
  • Good chemical resistance
Durable product has good temperature and chemical resistance.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403, Type 1 Class 4.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty